Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZW10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004724.3 | NP_004715.1 | 16 | 2906 | ||
XM_017018558.1 | XP_016874047.1 | 15 | 2654 | |||
XM_017018559.1 | XP_016874048.1 | 16 | 2862 | |||
GenBank mRNA | AK303871.1 | - | 15 | 2585 | ||
DB506745.2 | - | 2 | 613 | |||
U54996.1 | - | 16 | 2829 |