Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172947.3 | NP_766535.1 | 4, 5 | 537 | ||
GenBank mRNA | AK011126.1 | - | 4, 5 | 524 | ||
AK075668.1 | - | 4, 5 | 539 | |||
AK081685.1 | - | 512 | ||||
AK132889.1 | - | 4, 5 | 534 | |||
AK151118.1 | - | 540 | ||||
BC027784.1 | - | 4, 5 | 550 | 5343888 |