Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172947.3 | NP_766535.1 | 5 | 726 | ||
GenBank mRNA | AK011126.1 | - | 5 | 713 | ||
AK075668.1 | - | 5 | 728 | |||
AK081685.1 | - | 701 | ||||
AK151118.1 | - | 729 | ||||
BC027784.1 | - | 5 | 739 | 5343888 |