Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172951.3 | NP_766539.2 | 17 | 1770 | ||
GenBank mRNA | AK008673.1 | - | 17 | 1680 | ||
AK035310.1 | - | 17 | 1724 | |||
AK087560.1 | - | 17 | 1709 | |||
AK137345.1 | - | 17 | 1730 | |||
BC058204.1 | - | 16 | 1532 |