Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gphn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145965.2 | NP_666077.2 | 13, 14 | 1433 | ||
NM_172952.3 | NP_766540.2 | 14, 15 | 1424 | |||
GenBank mRNA | AK082353.1 | - | 14, 15 | 1346 | ||
BC027112.1 | - | 2, 3 | 220 | |||
BC127058.1 | - | 13, 14 | 1355 | 40109771 |