Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172953.3 | NP_766541.2 | 2 | 374 | ||
GenBank mRNA | AK077160.1 | - | 2 | 375 | ||
AK087714.1 | - | 2 | 337 | |||
BC090981.1 | - | 2 | 337 | 30285598 |