Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290786.1 | NP_001277715.1 | 4 | 1348 | ||
NM_172979.3 | NP_766567.1 | 4 | 1403 | |||
GenBank mRNA | AK028832.1 | - | 3 | 598 | ||
AK052840.1 | - | 4 | 1290 | |||
AK142590.1 | - | 4 | 1290 | |||
AY198336.1 | - | 4 | 1321 |