Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039866.1 | - | 16 | 1910 | ||
AK050894.1 | - | 19 | 2386 | |||
BC012520.1 | - | 7 | 1355 | |||
BC019659.1 | - | 10 | 1216 | |||
BC027074.1 | - | 3 | 632 | |||
BC050089.1 | - | 20 | 2525 | 30018732 |