Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256910.1 | NP_001243839.1 | 13 | 2240 | ||
NM_004728.3 | NP_004719.2 | 13 | 2121 | |||
XM_011540336.1 | XP_011538638.1 | 12 | 1951 | |||
XM_017016910.1 | XP_016872399.1 | 12 | 1869 | |||
GenBank mRNA | AB062394.1 | - | 5 | 600 | ||
AK025629.1 | - | 10 | 1423 | |||
AK315585.1 | - | 13 | 2070 | |||
BC008071.2 | - | 13 | 2059 | 2987980 | ||
BX648405.1 | - | 13 | 2007 | |||
CR749598.1 | - | 13 | 2241 | |||
U41387.1 | - | 13 | 2072 |