Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109749.1 | NP_001103219.1 | 12, 13 | 1580 | ||
NM_001109751.1 | NP_001103221.1 | 11, 12 | 1668 | |||
NM_173004.3 | NP_766592.2 | 12, 13 | 1580 | |||
GenBank mRNA | AK036262.1 | - | 12, 13 | 1550 | ||
AK043507.1 | - | 9, 10 | 1525 | |||
AK044694.1 | - | 2, 3 | 328 | |||
AK163520.1 | - | 9, 10 | 1373 | |||
AK173340.1 | - | 5, 6 | 699 | |||
BC046975.1 | - | 9, 10 | 1202 | |||
BC115766.1 | - | 12, 13 | 1470 | 40060288 |