Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pon3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173006.1 | NP_766594.1 | 2 | 94 | ||
GenBank mRNA | AK088572.1 | - | 2 | 94 | ||
BC099416.1 | - | 2 | 114 | 30847745 | ||
L76193.1 | - | 2 | 87 |