Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173011.2 | NP_766599.2 | 9, 10 | 1246 | ||
GenBank mRNA | AF212319.1 | - | 9, 10 | 1217 | ||
AK088110.1 | - | 9, 10 | 1218 | |||
AK145753.1 | - | 9, 10 | 1191 | |||
AK146564.1 | - | 9, 10 | 1332 | |||
AK161640.1 | - | 9, 10 | 1230 | |||
AK169395.1 | - | 9, 10 | 1243 | |||
BC060030.1 | - | 9, 10 | 1234 | 6515657 |