Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173030.2 | NP_766618.2 | 6 | 1112 | ||
GenBank mRNA | AB082928.1 | - | 4 | 600 | ||
AK038387.1 | - | 6 | 1113 | |||
AK045417.1 | - | 6 | 1113 | |||
BC131651.1 | - | 6 | 791 | 40110517 | ||
BC131652.1 | - | 5 | 822 | 40110518 |