Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173037.1 | NP_766625.1 | 6 | 1257 | ||
GenBank mRNA | AK054283.1 | - | 6 | 1211 | ||
AK076373.1 | - | 6 | 1180 | |||
AK077757.1 | - | 6 | 1243 | |||
AK085030.1 | - | 6 | 1257 | |||
AK173250.1 | - | 6 | 1164 | |||
BC066134.1 | - | 6 | 1187 | |||
BC066869.1 | - | 6 | 1187 | 30106406 | ||
BC069838.1 | - | 6 | 1187 | 30106422 |