Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbcel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173038.3 | NP_766626.1 | 3 | 411 | ||
GenBank mRNA | AK052261.1 | - | 3 | 380 | ||
AK077017.1 | - | 3 | 413 | |||
AK084854.1 | - | 3 | 424 | |||
BC047462.1 | - | 3 | 278 | |||
BC139386.1 | - | 3 | 409 | 8862408 | ||
BC139387.1 | - | 3 | 409 | 8862409 |