Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbcel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173038.3 | NP_766626.1 | 5 | 768 | ||
GenBank mRNA | AK052261.1 | - | 5 | 737 | ||
AK077017.1 | - | 5 | 770 | |||
AK084854.1 | - | 5 | 782 | |||
AK164693.1 | - | 1 | 1583 | |||
AK212783.1 | - | 2 | 266 | |||
BC047462.1 | - | 5 | 635 | |||
BC139386.1 | - | 5 | 766 | 8862408 | ||
BC139387.1 | - | 5 | 766 | 8862409 |