Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp775 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173429.2 | NP_775605.1 | 2 | 1850 | ||
GenBank mRNA | AK048054.1 | - | 2 | 1850 | ||
AK163249.1 | - | 3 | 1957 | |||
AK167452.1 | - | 3 | 1956 |