Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173739.3 | NP_776100.2 | 1 | 614 | ||
GenBank mRNA | AB175685.1 | - | 1 | 184 | ||
AK084671.1 | - | 1 | 619 | |||
BC024988.1 | - | 1 | 614 | 5044626 |