Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004752.3 | NP_004743.1 | 5 | 1592 | ||
GenBank mRNA | AF079550.1 | - | 5 | 1592 | ||
AF091149.1 | - | 5 | 1557 | |||
BC069603.1 | - | 5 | 1569 | 7262338 | ||
BC117316.1 | - | 5 | 1592 | 40125867 | ||
BC117318.1 | - | 5 | 1592 | 40125869 |