Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173781.4 | NP_776142.1 | 8 | 1376 | ||
GenBank mRNA | AK035893.1 | - | 8 | 1335 | ||
AK082732.1 | - | 8 | 1501 | |||
BC059903.1 | - | 4 | 959 | |||
BC060618.1 | - | 8 | 1377 | 6406134 |