Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290801.1 | NP_001277730.1 | 3 | 529 | ||
NM_173870.3 | NP_776295.1 | 4 | 608 | |||
GenBank mRNA | AB053217.1 | - | 3 | 284 | ||
AK052278.1 | - | 1 | 105 | |||
AK155724.1 | - | 4 | 609 | |||
AK163327.1 | - | 3 | 361 |