Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175483.5 | NP_780692.2 | 1 | 1911 | ||
GenBank mRNA | AK048566.1 | - | 1 | 416 | ||
AK053761.1 | - | 1 | 446 | |||
AK142274.1 | - | 1 | 1912 | |||
BC096472.1 | - | 1 | 661 | 3501525 |