Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175488.6 | NP_780697.2 | 13 | 1387 | ||
GenBank mRNA | AK029803.1 | - | 13 | 1388 | ||
AK030198.1 | - | 13 | 1376 | |||
AK030205.1 | - | 10 | 1037 |