Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177673.1 | NP_001171144.1 | 3 | 2288 | ||
NM_001177674.1 | NP_001171145.1 | 3 | 2470 | |||
NM_175493.4 | NP_780702.1 | 3 | 2591 | |||
GenBank mRNA | AK036659.1 | - | 2 | 2300 | ||
AK154649.1 | - | 2 | 2263 | |||
AK155886.1 | - | 2 | 2280 |
Set Membership: |
G-protein coupled receptor |