Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NREP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142474.1 | NP_001135946.1 | 4 | 557 | ||
NM_001142475.1 | NP_001135947.1 | 4 | 587 | |||
NM_001142476.1 | NP_001135948.1 | 5 | 639 | |||
NM_001142477.1 | NP_001135949.1 | 5 | 530 | |||
NM_001142478.1 | NP_001135950.1 | 5 | 506 | |||
NM_001142479.1 | NP_001135951.1 | 5 | 543 | |||
NM_001142480.1 | NP_001135952.1 | 5 | 537 | |||
NM_001142481.1 | NP_001135953.1 | 5 | 446 | |||
NM_001142482.1 | NP_001135954.1 | 5 | 453 | |||
NM_001142483.1 | NP_001135955.1 | 3 | 742 | |||
NM_004772.2 | NP_004763.1 | 4 | 434 | |||
XM_006714732.1 | XP_006714795.1 | 3 | 354 | |||
XM_006714733.2 | XP_006714796.1 | 4 | 335 | |||
GenBank mRNA | AF119859.1 | - | 1 | 234 | ||
AK298779.1 | - | 4 | 482 | |||
AK309667.1 | - | 3 | 265 | |||
AL547663.3 | - | 5 | 535 | |||
BC011050.1 | - | 5 | 467 | 4340005 | ||
BC019068.2 | - | 4 | 370 | 4893075 | ||
BC072013.1 | - | 4 | 393 | 5922876 | ||
BC072443.1 | - | 4 | 398 | 6162335 | ||
DA027062.1 | - | 4 | 557 | |||
DA078027.1 | - | 3 | 467 | |||
DA520221.1 | - | 5 | 465 | |||
DA683910.1 | - | 5 | 453 | |||
DA694580.1 | - | 4 | 452 | |||
DA744980.1 | - | 5 | 390 | |||
DA752769.1 | - | 5 | 446 | |||
DA908653.1 | - | 4 | 434 | |||
DA939543.1 | - | 5 | 530 | |||
U30521.1 | - | 5 | 465 | |||
U36189.1 | - | 3 | 278 |