Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nyap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175521.3 | NP_780730.2 | 3 | 1309 | ||
GenBank mRNA | AB429289.1 | - | 3 | 1309 | ||
AK032567.1 | - | 1 | 482 | |||
AK080821.1 | - | 4 | 1679 | |||
AK141276.1 | - | 4 | 1166 | |||
AK148980.1 | - | 1 | 273 | |||
BC057372.1 | - | 3 | 1309 | 5686602 |