Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271755.1 | NP_001258684.1 | 10 | 1651 | ||
NM_001271756.1 | NP_001258685.1 | 10 | 1290 | |||
NM_004791.2 | NP_004782.1 | 11 | 1798 | |||
GenBank mRNA | AB008375.1 | - | 12 | 1662 | ||
AF072752.1 | - | 11 | 1776 | |||
AK095875.1 | - | 11 | 1438 | |||
AK298571.1 | - | 10 | 1290 | |||
AK308194.1 | - | 10 | 1568 | |||
BC036788.1 | - | 11 | 1719 | 5753239 |