Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175548.3 | NP_780757.1 | 6 | 1770 | ||
GenBank mRNA | AK044845.1 | - | 6 | 1770 |