Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Robo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175549.4 | NP_780758.3 | 3, 4 | 539 | ||
GenBank mRNA | AK031495.1 | - | 3, 4 | 1065 | ||
AK035110.1 | - | 3, 4 | 1413 | |||
AK085461.1 | - | 3, 4 | 1164 | |||
BC055333.1 | - | 3, 4 | 847 | 6409283 | ||
DQ533875.1 | - | 4, 5 | 819 | |||
DQ533876.1 | - | 3, 4 | 539 |