Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175552.4 | NP_780761.1 | 27 | 2828 | ||
GenBank mRNA | AK050738.1 | - | 27 | 2796 | ||
AK050797.1 | - | 27 | 2799 | |||
BC050990.1 | - | 18 | 1757 | |||
BC063100.1 | - | 27 | 2814 | 6332749 |