Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp13a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284375.1 | NP_001271304.1 | 2 | 347 | ||
NM_175650.4 | NP_783581.2 | 2 | 347 | |||
GenBank mRNA | AK083102.1 | - | 2 | 322 | ||
AK083121.1 | - | 2 | 348 | |||
AK158355.1 | - | 2 | 167 | |||
BC115739.1 | - | 2 | 282 | 40086065 | ||
CB524474.1 | - | 2 | 149 |