Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp13a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284375.1 | NP_001271304.1 | 3, 4 | 575 | ||
NM_175650.4 | NP_783581.2 | 3, 4 | 575 | |||
GenBank mRNA | AK083102.1 | - | 3, 4 | 550 | ||
AK083121.1 | - | 3, 4 | 576 | |||
AK158355.1 | - | 3, 4 | 395 | |||
BC115739.1 | - | 3, 4 | 510 | 40086065 | ||
CB524474.1 | - | 3, 4 | 377 |