Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim12c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146007.1 | NP_001139479.1 | 5, 6 | 998 | ||
NM_175677.4 | NP_783608.3 | 5, 6 | 879 | |||
GenBank mRNA | AK033764.1 | - | 5, 6 | 999 | ||
AK050485.1 | - | 5, 6 | 1003 | |||
AK146437.1 | - | 5, 6 | 958 | |||
BC038937.1 | - | 5, 6 | 879 | 2648191 |
Gene Symbol: Trim12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023835.2 | NP_076324.2 | 5, 6 | 918 | ||
GenBank mRNA | AK009769.1 | - | 5, 6 | 880 | ||
BC094899.1 | - | 4, 5 | 782 | 30041200 |