Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim12c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146007.1 | NP_001139479.1 | 6 | 1031 | ||
NM_175677.4 | NP_783608.3 | 6 | 912 | |||
GenBank mRNA | AK033764.1 | - | 6 | 1032 | ||
AK050485.1 | - | 6 | 1036 | |||
AK146437.1 | - | 6 | 991 | |||
BC038937.1 | - | 6 | 912 | 2648191 |
Gene Symbol: Trim12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023835.2 | NP_076324.2 | 6 | 951 | ||
GenBank mRNA | AK009769.1 | - | 6 | 913 | ||
BC094899.1 | - | 5 | 815 | 30041200 |