Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim12c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146007.1 | NP_001139479.1 | 8 | 1781 | ||
NM_175677.4 | NP_783608.3 | 8 | 1662 | |||
GenBank mRNA | AK033764.1 | - | 8 | 1781 | ||
AK146437.1 | - | 8 | 1741 | |||
BC038937.1 | - | 8 | 1662 | 2648191 |