Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175678.3 | NP_783609.1 | 9 | 961 | ||
GenBank mRNA | AK033957.1 | - | 9 | 962 | ||
AK041451.1 | - | 8 | 860 | |||
AK041501.1 | - | 3 | 257 | |||
AY299387.1 | - | 8 | 897 |
Set Membership: |
G-protein coupled receptor |