Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287489.1 | NP_001274418.1 | 46 | 6522 | ||
NM_004802.3 | NP_004793.2 | 30 | 4518 | |||
NM_194248.2 | NP_919224.1 | 47 | 6720 | |||
NM_194322.2 | NP_919303.1 | 29 | 4672 | |||
NM_194323.2 | NP_919304.1 | 29 | 4320 | |||
GenBank mRNA | AF107403.1 | - | 30 | 4518 | ||
AF183185.1 | - | 47 | 6720 | |||
AF183186.1 | - | 29 | 4672 | |||
AF183187.1 | - | 29 | 4320 |