Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146099.1 | NP_001139571.1 | 3 | 531 | ||
NM_002498.2 | NP_002489.1 | 3 | 531 | |||
NM_152720.2 | NP_689933.1 | 3 | 550 | |||
NR_027415.2 | 3 | 531 | ||||
GenBank mRNA | AB072828.1 | - | 2 | 200 | ||
AK131359.1 | - | 2 | 1449 | |||
AK290259.1 | - | 3 | 263 | |||
AL833951.1 | - | 3 | 390 | |||
BC019916.1 | - | 3 | 258 | 5000918 | ||
BQ648157.1 | - | 3 | 467 | |||
CV029372.1 | - | 2 | 155 | |||
DB185803.1 | - | 3 | 528 |