Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170742.1 | NP_001164213.1 | 3 | 338 | ||
NM_001170743.1 | NP_001164214.1 | 3 | 338 | |||
NM_176923.4 | NP_795897.2 | 3 | 338 | |||
GenBank mRNA | AK030644.1 | - | 3 | 219 | ||
AK170910.1 | - | 3 | 337 | |||
AK173301.1 | - | 3 | 301 | |||
BC066082.1 | - | 3 | 323 | 30355918 | ||
EF011622.1 | - | 3 | 323 |