Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143995.2 | NP_001137467.1 | 9 | 1512 | ||
NM_001307951.1 | NP_001294880.1 | 8 | 1498 | |||
NM_004811.2 | NP_004802.1 | 9 | 1449 | |||
XM_006718750.1 | XP_006718813.1 | 9 | 1545 | |||
XM_011545388.2 | XP_011543690.1 | 8 | 2169 | |||
XM_011545390.2 | XP_011543692.1 | 8 | 2479 | |||
XM_011545391.2 | XP_011543693.1 | 8 | 1445 | |||
GenBank mRNA | AF062075.1 | - | 9 | 1397 | ||
AK223165.1 | - | 9 | 1388 | |||
AK299264.1 | - | 8 | 1479 | |||
BC019035.2 | - | 9 | 1371 | 4575069 |