Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004815.3 | NP_004806.3 | 21, 22 | 3170 | ||
XM_006711048.3 | XP_006711111.1 | 20, 21 | 2804 | |||
XM_011542438.2 | XP_011540740.1 | 21, 22 | 3045 | |||
XM_011542439.2 | XP_011540741.1 | 21, 22 | 2890 | |||
XM_011542440.2 | XP_011540742.1 | 20, 21 | 2764 | |||
GenBank mRNA | AB208873.1 | - | 20, 21 | 2776 | ||
AK310491.1 | - | 21, 22 | 2890 | |||
AL833445.1 | - | 9, 10 | 7379 | |||
BC093741.1 | - | 20, 21 | 2843 | 7939586 | ||
BC093767.1 | - | 20, 21 | 2846 | 7939612 | ||
CB852768.1 | - | 2, 3 | 378 | |||
U90920.1 | - | 21, 22 | 2984 |