Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176940.5 | NP_795914.3 | 5 | 1086 | ||
GenBank mRNA | AK034868.1 | - | 785 | |||
AK038792.1 | - | 4 | 778 | |||
AK162888.1 | - | 5 | 1081 | |||
BC082552.1 | - | 5 | 1026 | |||
BC144786.1 | - | 3 | 676 | 9053334 | ||
BC144787.1 | - | 3 | 676 | |||
BC145703.1 | - | 3 | 676 | 40131006 |