Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM189A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127608.1 | NP_001121080.1 | 11 | 2031 | ||
NM_004816.3 | NP_004807.3 | 11 | 2347 | |||
XM_005252307.4 | XP_005252364.1 | 11 | 2908 | |||
XM_017015324.1 | XP_016870813.1 | 10 | 2092 | |||
XM_017015325.1 | XP_016870814.1 | 10 | 1738 | |||
GenBank mRNA | AK056152.1 | - | 9 | 1975 | ||
AK126127.1 | - | 11 | 2348 | |||
BC021685.1 | - | 7 | 1468 | |||
BX641153.1 | - | 7 | 9413 |