Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176950.3 | NP_795924.2 | 2 | 620 | ||
GenBank mRNA | AK079000.1 | - | 2 | 621 | ||
DQ012031.1 | - | 2 | 620 |