Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xkr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113350.2 | NP_001106821.1 | 5 | 728 | ||
NM_001286469.1 | NP_001273398.1 | 5 | 1801 | |||
NM_176951.5 | NP_795925.1 | 5 | 1068 | |||
GenBank mRNA | AK030692.1 | - | 5 | 1053 | ||
AK045098.1 | - | 5 | 1047 | |||
AY534251.1 | - | 5 | 728 | |||
BC098477.1 | - | 5 | 1801 | |||
BC117838.1 | - | 4 | 635 | 40093151 | ||
BC117839.1 | - | 4 | 635 | 40093152 |