Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037277.1 | NP_001032354.1 | 3, 4 | 221 | ||
NR_036605.1 | 2, 3 | 292 | ||||
GenBank mRNA | AB016043.1 | - | 3, 4 | 306 | ||
AB017971.1 | - | 2, 3 | 137 | |||
AB019036.1 | - | 3, 4 | 347 | |||
AF057698.1 | - | 3, 4 | 375 | |||
AF125394.1 | - | 3, 4 | 284 | |||
AK025139.1 | - | 3, 4 | 369 | |||
AK293278.1 | - | 2, 3 | 275 | |||
BC005252.1 | - | 3, 4 | 369 | 4051481 | ||
BC067768.1 | - | 3, 4 | 309 | 5295932 | ||
BI462253.1 | - | 3, 4 | 396 | |||
BI520491.1 | - | 3, 4 | 224 | |||
BI963107.1 | - | 2, 3 | 242 | |||
CB216084.1 | - | 3, 4 | 387 | |||
DB473584.1 | - | 3, 4 | 397 |