Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tigar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177003.5 | NP_795977.1 | 6 | 1345 | ||
GenBank mRNA | AK036082.1 | - | 6 | 1336 | ||
AK145896.1 | - | 6 | 1330 |