Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018009.3 | NP_001018009.2 | 9 | 1430 | ||
NM_004844.4 | NP_004835.2 | 9 | 1549 | |||
XM_011534251.2 | XP_011532553.1 | 9 | 1361 | |||
XM_011534253.1 | XP_011532555.1 | 8 | 1249 | |||
XM_017007521.1 | XP_016863010.1 | 9 | 1344 | |||
XM_017007522.1 | XP_016863011.1 | 10 | 1987 | |||
XM_017007523.1 | XP_016863012.1 | 9 | 1765 | |||
XM_017007524.1 | XP_016863013.1 | 9 | 1297 | |||
XM_017007525.1 | XP_016863014.1 | 9 | 1211 | |||
GenBank mRNA | AB005047.1 | - | 9 | 1301 | ||
AK090524.1 | - | 11 | 1653 | |||
AK313167.1 | - | 9 | 1507 | |||
AL117422.1 | - | 9 | 1505 | |||
BC006169.1 | - | 9 | 1404 | |||
BC010123.2 | - | 9 | 1480 | 3841593 | ||
BC023195.1 | - | 9 | 1221 | |||
BX641159.1 | - | 2 | 1480 |