Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cobll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027225.1 | NP_081501.1 | 11 | 2821 | ||
NM_177025.5 | NP_795999.2 | 12 | 2932 | |||
GenBank mRNA | AK037010.1 | - | 13 | 2926 | ||
AK052533.1 | - | 11 | 2770 | |||
AK135859.1 | - | 3 | 1160 | |||
AK144943.1 | - | 13 | 2979 | |||
AK173076.1 | - | 11 | 2652 | |||
AY308746.1 | - | 12 | 2932 | |||
BC067007.1 | - | 11 | 2818 | 5702930 |