Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177036.5 | NP_796010.1 | 7 | 1055 | ||
GenBank mRNA | AK047942.1 | - | 7 | 1042 | ||
AK079290.1 | - | 7 | 1056 | |||
AK163794.1 | - | 7 | 1043 | |||
BC107227.1 | - | 7 | 983 | 40051960 | ||
BC107228.2 | - | 7 | 983 | 40051962 | ||
BC109333.3 | - | 7 | 983 | 40051723 |